Skip to Content
Merck
All Photos(1)

Key Documents

698466

Sigma-Aldrich

Triisopropylphosphonium tetrafluoroborate

97%

Synonym(s):

Tris(1-methylethyl)phosphine tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H22BF4P
CAS Number:
Molecular Weight:
248.05
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

functional group

phosphine

SMILES string

F[B-](F)(F)F.CC(C)[PH+](C(C)C)C(C)C

InChI

1S/C9H21P.BF4/c1-7(2)10(8(3)4)9(5)6;2-1(3,4)5/h7-9H,1-6H3;/q;-1/p+1

InChI key

KGBIZABAOCDZNU-UHFFFAOYSA-O

Application

Reactant for:
  • Synthesis of triisopropylphosphine complexes of ruthenium(II)
  • Imine hydrogenation
  • Dihydride and alkene-η6-arene complexes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service