Skip to Content
Merck
All Photos(2)

Key Documents

630063

Sigma-Aldrich

Aluminum chloride, functionalized silica gel

40-63 μm, extent of labeling: 1.5 mmol/g loading

Synonym(s):

Aluminum trichloride, functionalized silica gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352311
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

core: aluminum
reagent type: Lewis acid
reagent type: catalyst

Quality Level

extent of labeling

1.5 mmol/g loading

surface area

500 m2/g

particle size

40-63 μm

pore size

60 Å pore size

Application

Silica-bound Lewis acid. Catalyst for Friedel-Crafts reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service