616699
Mevalonolactone-(methyl-d3)
99 atom % D, ≥97% (CP)
Synonym(s):
DL-Mevalonic-methyl-d3 lactone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6D3H7O3
Molecular Weight:
133.16
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % D
Quality Level
Assay
≥97% (CP)
form
solid
bp
150 °C/5 mmHg (lit.)
mp
28 °C (lit.)
mass shift
M+3
SMILES string
[2H]C([2H])([2H])C1(O)CCOC(=O)C1
InChI
1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i1D3
InChI key
JYVXNLLUYHCIIH-FIBGUPNXSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service