Skip to Content
Merck
All Photos(1)

Key Documents

606715

Sigma-Aldrich

Lindane-13C6 (γ-BHC)

99 atom % 13C

Synonym(s):

1,2,3,4,5,6-Hexachloro cyclohexane-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H6Cl6
CAS Number:
Molecular Weight:
296.79
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

mp

113-115 °C (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+6

SMILES string

Cl[13C@@H]1[13C@@H](Cl)[13C@@H](Cl)[13C@H](Cl)[13C@@H](Cl)[13C@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5+,6+/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

JLYXXMFPNIAWKQ-BDHSFOLOSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service