Skip to Content
Merck
All Photos(1)

Key Documents

600202

Sigma-Aldrich

Barium carbonate-13C

90 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba13CO3
CAS Number:
Molecular Weight:
198.33
Beilstein:
4826209
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:

isotopic purity

90 atom % 13C

mol wt

198.23 by atom % calculation

mp

811 °C (lit.)

mass shift

M+1

SMILES string

[Ba++].[O-][13C]([O-])=O

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2/i1+1;

InChI key

AYJRCSIUFZENHW-YTBWXGASSA-L

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service