597430
4-Ethynylbenzaldehyde
97%
Synonym(s):
4-Formylphenylacetylene, p-Ethynylbenzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
mp
89-93 °C (lit.)
SMILES string
O=Cc1ccc(cc1)C#C
InChI
1S/C9H6O/c1-2-8-3-5-9(7-10)6-4-8/h1,3-7H
InChI key
BGMHQBQFJYJLBP-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of chromatography. A, 1355, 206-210 (2014-07-09)
A novel pre-column fluorescence derivatization method for chromatographic analysis of azide compounds was developed based on the Huisgen reaction, which is a specific cycloaddition reaction between an alkyne and an azide. We designed and synthesized a fluorescent alkyne, 2-(4-ethynylphenyl)-4,5-diphenyl-1H-imidazole (DIB-ET)
ACS nano, 8(5), 5240-5248 (2014-04-18)
Simple preparation methods of chemically versatile and highly functionalizable surfaces remain rare and present a challenging research objective. Here, we demonstrate a simultaneous electropolymerization and electro-click functionalization process (SEEC) for one-pot self-construction of aniline- and naphthalene-based functional polymer films where
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service