586625
Acrylic acid-13C3
≥99 atom % 13C, ≥99% (CP), contains hydroquinone as stabilizer
Synonym(s):
13C Labeled acrylic acid
About This Item
Recommended Products
isotopic purity
≥99 atom % 13C
Quality Level
Assay
≥99% (CP)
contains
hydroquinone as stabilizer
impurities
≤5% water
bp
139 °C (lit.)
mp
13 °C (lit.)
density
1.094 g/mL at 25 °C
mass shift
M+3
storage temp.
2-8°C
SMILES string
O[13C](=O)[13CH]=[13CH2]
InChI
1S/C3H4O2/c1-2-3(4)5/h2H,1H2,(H,4,5)/i1+1,2+1,3+1
InChI key
NIXOWILDQLNWCW-VMIGTVKRSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
114.8 °F - closed cup
Flash Point(C)
46 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service