All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4N2O4 · 2H2O
CAS Number:
Molecular Weight:
204.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
96%
mp
272-277 °C (lit.)
functional group
carboxylic acid
SMILES string
[H]O[H].[H]O[H].OC(=O)c1cnc(cn1)C(O)=O
InChI
1S/C6H4N2O4.2H2O/c9-5(10)3-1-7-4(2-8-3)6(11)12;;/h1-2H,(H,9,10)(H,11,12);2*1H2
InChI key
GVEVNOSZLMMBOR-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service