Skip to Content
Merck
All Photos(1)

Key Documents

52045

Sigma-Aldrich

Hexacyclen

≥97.0% (NT)

Synonym(s):

1,4,7,10,13,16-Hexaazacyclooctadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H30N6
CAS Number:
Molecular Weight:
258.41
Beilstein:
1105038
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (NT)

form

solid

mp

147-152 °C

SMILES string

C1CNCCNCCNCCNCCNCCN1

InChI

1S/C12H30N6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h13-18H,1-12H2

InChI key

RVJABZUDCPZPPM-UHFFFAOYSA-N

Other Notes

Nitrogen analog of 18-crown-6 in which the secondary amines act as excellent electron donors to transition metal cations. Review

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.H. Busch
Accounts of Chemical Research, 11, 392-392 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service