Skip to Content
Merck
All Photos(1)

Key Documents

489190

Sigma-Aldrich

Eicosane-d42

98 atom % D, 98% (CP)

Synonym(s):

Deuterated eicosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)18CD3
CAS Number:
Molecular Weight:
324.81
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solid

bp

220 °C/30 mmHg (lit.)

mp

36-38 °C (lit.)

mass shift

M+42

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C20H42/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3-20H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2

InChI key

CBFCDTFDPHXCNY-ILDSYVRBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruixin Xu et al.
The Science of the total environment, 720, 137470-137470 (2020-04-25)
Particulate matter originated from traffic has attracted major interest over the last few years. The semi-volatile organic component of the particles may evaporate with dispersion away from the emission source, creating vapour which may oxidise to form secondary organic aerosol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service