Skip to Content
Merck
All Photos(1)

Key Documents

47751

Sigma-Aldrich

Fmoc-Tyr-OH

≥97.0% (HPLC)

Synonym(s):

Nα-Fmoc-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21NO5
CAS Number:
Molecular Weight:
403.43
Beilstein:
3573123
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥97.0% (HPLC)

optical activity

[α]20/D −22±2°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H21NO5/c26-16-11-9-15(10-12-16)13-22(23(27)28)25-24(29)30-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22,26H,13-14H2,(H,25,29)(H,27,28)/t22-/m0/s1

InChI key

SWZCTMTWRHEBIN-QFIPXVFZSA-N

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chiao-Ting Yen et al.
European journal of medicinal chemistry, 44(5), 1933-1940 (2008-12-27)
Twenty-four new dipeptide analogs (1-24) of aurantiamide acetate were designed, synthesized, and assayed for effects on superoxide anion generation and elastase release by human neutrophils in response to fMLP/CB. Among them, seven N-(fluorenyl-9-methoxycarbonyl) (Fmoc)-dipeptides (6, 9, 12, 14, 17, 18
Weidang Wu et al.
Cancer science, 106(6), 747-756 (2015-04-14)
L-type amino acid transporter 1 (LAT1), overexpressed on the membrane of various tumor cells, is a potential target for tumor-targeting therapy. This study aimed to develop a LAT1-mediated chemotherapeutic agent. We screened doxorubicin modified by seven different large neutral amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service