All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H8N2O
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
mp
153-155 °C (lit.)
functional group
aldehyde
SMILES string
O=Cc1ccc(cc1)-n2ccnc2
InChI
1S/C10H8N2O/c13-7-9-1-3-10(4-2-9)12-6-5-11-8-12/h1-8H
InChI key
DCICUQFMCRPKHZ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jakub Hrubý et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-04)
Studying the properties of complex molecules on surfaces is still mostly an unexplored research area because the deposition of the metal complexes has many pitfalls. Herein, we probed the possibility to produce surface hybrids by depositing a Co(II)-based complex with
Vijay K Marrapu et al.
European journal of medicinal chemistry, 46(9), 4302-4310 (2011-07-19)
A series of twenty seven novel aryloxy azolyl chalcones were synthesized and evaluated in vitro for the growth inhibition of Mycobacterium tuberculosis H37Rv. Ten compounds from this series exhibited good activity with MIC in the range of 3.12-0.78 μg/mL and
Tanvir Hussain et al.
European journal of medicinal chemistry, 44(11), 4654-4660 (2009-08-12)
A series of new 3-[4-(1H-imidazol-1-yl) phenyl]prop-2-en-1-ones were synthesized by the condensation of various acetophenones with 4-(1H-imidazol-1-yl) benzaldehyde which was itself prepared by the N-arylation of imidazole using hexadecyltrimethylammonium bromide as catalyst for the first time. All the synthesized compounds were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service