438200
Poly(ethylene glycol-ran-propylene glycol)
average Mn ~12,000
Synonym(s):
PEG/PPG Copolymer
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
form
viscous liquid
Quality Level
mol wt
average Mn ~12,000
refractive index
n20/D 1.466
viscosity
35,000 cSt(25 °C)(lit.)
bp
>200 °C (lit.)
transition temp
pour point 4 °C
solubility
H2O: soluble at <70 °C
density
1.092 g/mL at 25 °C
SMILES string
OCCO.CC(O)CO
InChI key
HQPMKSGTIOYHJT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Hydraulic fluids, metal working fluids and lubricants, heat transfer fluids, solder assist fluids, quenchants, lubricants, solvents, plasticizers and foam control agents.
Features and Benefits
Good lubricity under load, noncorrosive to metals. Very low ash content, low pour point, excellent thermal stability.
Physical form
Dihydroxy functional.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service