425222
Ethyl 2-(bromomethyl)acrylate
98%
Synonym(s):
2-(Bromomethyl)acrylic acid ethyl ester, Ethyl α-(bromomethyl)acrylate, Ethyl 2-(bromomethyl)-2-propenoate, Ethyl 2-(bromomethyl)propenoate
About This Item
Recommended Products
Assay
98%
refractive index
n20/D 1.479 (lit.)
bp
38 °C/0.8 mmHg (lit.)
density
1.398 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
[H]\C([H])=C(\CBr)C(=O)OCC
InChI
1S/C6H9BrO2/c1-3-9-6(8)5(2)4-7/h2-4H2,1H3
InChI key
MTCMFVTVXAOHNQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
177.8 °F - closed cup
Flash Point(C)
81 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service