423793
Triisobutylaluminum solution
1.0 M in hexanes
Synonym(s):
TIBA solution
About This Item
Recommended Products
form
liquid
Quality Level
concentration
1.0 M in hexanes
bp
68-69 °C
density
0.695 g/mL at 25 °C
SMILES string
CC(C)C[Al](CC(C)C)CC(C)C
InChI
1S/3C4H9.Al/c3*1-4(2)3;/h3*4H,1H2,2-3H3;
InChI key
MCULRUJILOGHCJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Signal Word
Danger
Hazard Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1
Target Organs
Central nervous system, Nervous system
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service