Skip to Content
Merck
All Photos(1)

Key Documents

412074

Sigma-Aldrich

5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine

Dye content 95 %

Synonym(s):

5,9,14,18,23,27,32,36-Octabutoxynaphthalocyanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H90N8O8
CAS Number:
Molecular Weight:
1291.62
Beilstein:
4949121
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 95%

Quality Level

λmax

867 nm

SMILES string

CCCCOc1c2ccccc2c(OCCCC)c3c4nc(nc5[nH]c(nc6nc(nc7[nH]c(n4)c8c(OCCCC)c9ccccc9c(OCCCC)c78)c%10c(OCCCC)c%11ccccc%11c(OCCCC)c6%10)c%12c(OCCCC)c%13ccccc%13c(OCCCC)c5%12)c13

InChI

1S/C80H90N8O8/c1-9-17-41-89-65-49-33-25-26-34-50(49)66(90-42-18-10-2)58-57(65)73-81-74(58)86-76-61-62(70(94-46-22-14-6)54-38-30-29-37-53(54)69(61)93-45-21-13-5)78(83-76)88-80-64-63(71(95-47-23-15-7)55-39-31-32-40-56(55)72(64)96-48-24-16-8)79(84-80)87-77-60-59(75(82-77)85-73)67(91-43-19-11-3)51-35-27-28-36-52(51)68(60)92-44-20-12-4/h25-40H,9-24,41-48H2,1-8H3,(H2,81,82,83,84,85,86,87,88)

InChI key

BFXKASLQHBYWIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service