389927
Silicon 2,3-naphthalocyanine dichloride
Dye content 85 %
Synonym(s):
2,3-Naphthalocyaninato-dichlorosilane, Dichloro-2,3-naphthalocyaninatosilane
About This Item
Recommended Products
form
powder
Quality Level
composition
Dye content, 85%
λmax
780 nm
SMILES string
Cl[Si]1(Cl)n2c3nc4nc(nc5n1c(nc6nc(nc2c7cc8ccccc8cc37)c9cc%10ccccc%10cc69)c%11cc%12ccccc%12cc5%11)c%13cc%14ccccc%14cc4%13
InChI
1S/C48H24Cl2N8Si/c49-59(50)57-45-37-21-29-13-5-6-14-30(29)22-38(37)47(57)55-43-35-19-27-11-3-4-12-28(27)20-36(35)44(52-43)56-48-40-24-32-16-8-7-15-31(32)23-39(40)46(58(48)59)54-42-34-18-26-10-2-1-9-25(26)17-33(34)41(51-42)53-45/h1-24H/b53-41-,53-45-,54-42-,54-46-,55-43-,55-47-,56-44-,56-48-
InChI key
XIINLOVCJKUOQC-IIZSCGODSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service