Skip to Content
Merck
All Photos(1)

Key Documents

388211

Sigma-Aldrich

Perfluorooctanes

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

form

liquid

Quality Level

refractive index

n20/D 1.3 (lit.)

bp

103-105 °C (lit.)

density

1.766 g/mL at 25 °C (lit.)

Application

Perfluorooctanes can be used as a fluorous solvent for aerobic oxidation of alcohols to carbonyl compounds in fluorous biphasic systems. It can also be employed in the synthesis of perfluoroalkyl-substituted bis(oxazolines).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Perfluoroalkyl-Substituted Bis (oxazolines) as Ligands for Catalytic Enantioselective Reactions.
Annunziata R, et al.
European Journal of Organic Chemistry, 2003(7), 1191-1197 (2003)
Cobalt (II) perfluorooctane sulfonate catalyzed highly efficient aerobic oxidation of alcohols in fluorous biphasic systems.
Yi WB and Cai C
Australian Journal of Chemistry, 59(5), 349-353 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service