Skip to Content
Merck
All Photos(1)

Key Documents

377015

Sigma-Aldrich

(Chloromethyl)triphenylphosphonium chloride

97%

Synonym(s):

NSC 93980

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2P(Cl)(C6H5)3
CAS Number:
Molecular Weight:
347.22
Beilstein:
3599784
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

mp

262-264 °C (lit.)

SMILES string

[Cl-].ClC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H17ClP.ClH/c20-16-21(17-10-4-1-5-11-17,18-12-6-2-7-13-18)19-14-8-3-9-15-19;/h1-15H,16H2;1H/q+1;/p-1

InChI key

SXYFAZGVNNYGJQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used for corrosion inhibition of iron

Reactant involved in:
  • Acting as a potential electrophile
  • Elimination reactions for the synthesis of linear alkynes
  • Intramolecular nucleophilic vinylic substitution
  • Homologation of aldehydes
  • Endo-selective alkynol cycloisomerization

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service