Skip to Content
Merck
All Photos(1)

Key Documents

342017

Sigma-Aldrich

Heptafluoro-2-iodopropane

98%

Synonym(s):

2-Iodoperfluoropropane, Heptafluoroisopropyl iodide, Perfluoroisopropyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CFI
CAS Number:
Molecular Weight:
295.93
Beilstein:
1841228
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

7.12 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.329 (lit.)

bp

40 °C (lit.)

density

2.08 g/mL at 25 °C (lit.)

functional group

fluoro
iodo

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(I)C(F)(F)F

InChI

1S/C3F7I/c4-1(11,2(5,6)7)3(8,9)10

InChI key

BBZVTTKMXRPMHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptafluoro-2-iodopropane is a secondary fluoro alkyl iodide. It can be prepared by addition of iodine monofluoride to hexafluoropropene.

Application

Heptafluoro-2-iodopropane may be used in the preparation of perfluoropropyl magnesium bromide and perfluoropropyl magnesium iodide. It may be used as chain-transfer agent in telomerization reactions of:
  • 1,1-difluoroethylene and tetrafluoroethylene
  • CF3I, C2F5I and n-C3F7I with 1,1-difluoroethylene
  • n-C3F7I with tetrafluoroethylene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Telomerization reactions in the synthesis of models for some fluorocarbon Polymers.
Chambers RD, et al.
Tetrahedron, 20(3), 497-506 (1964)
372. Organometallic and metalloid compounds made from heptafluoro-2-iodopropane, and their properties.
Chambers RD, et al.
Journal of the Chemical Society, 1962, 1993-1999 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service