Skip to Content
Merck
All Photos(1)

Key Documents

333530

Sigma-Aldrich

2-Bromoallyltrimethylsilane

technical grade, 90%

Synonym(s):

2-Bromo-3-(trimethylsilyl)propene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(Br)CH2Si(CH3)3
CAS Number:
Molecular Weight:
193.16
Beilstein:
3600686
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

82-85 °C/60 mmHg (lit.)

density

1.121 g/mL at 25 °C (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

C[Si](C)(C)CC(Br)=C

InChI

1S/C6H13BrSi/c1-6(7)5-8(2,3)4/h1,5H2,2-4H3

InChI key

LUPQCAARZVEFMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service