332518
Dibenzo-30-crown-10
98%
Synonym(s):
2,3,17,18-Dibenzo-1,4,7,10,13,16,19,22,25,28-decaoxacyclotriaconta-2,17-diene
About This Item
Recommended Products
Quality Level
Assay
98%
form
solid
mp
106-108 °C (lit.)
SMILES string
C1COCCOc2ccccc2OCCOCCOCCOCCOc3ccccc3OCCOCCO1
InChI
1S/C28H40O10/c1-2-6-26-25(5-1)35-21-17-31-13-9-29-11-15-33-19-23-37-27-7-3-4-8-28(27)38-24-20-34-16-12-30-10-14-32-18-22-36-26/h1-8H,9-24H2
InChI key
MXCSCGGRLMRZMF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service