325538
Dysprosium(III) acetate hydrate
99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3CO2)3Dy · 4H2O
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.9% trace metals basis
form
powder
composition
Degree of hydration, ≤4
reaction suitability
core: dysprosium
reagent type: catalyst
SMILES string
CC([O-])=O.[Dy+3]
InChI
1S/3C2H4O2.Dy.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3
InChI key
AAGMOWMTSATFBV-UHFFFAOYSA-K
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Xixi Wang et al.
Nanoscale, 12(36), 18807-18814 (2020-09-25)
Mechanistic studies on photon upconversion play a critical role in the fundamental research of the luminescence of rare earth ions as well as their emerging applications. Energy migration mediated upconversion (EMU) has recently shown to be an important model for
Bo Zhou et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(3), 1700667-1700667 (2018-03-30)
Upconverting materials have achieved great progress in recent years, however, it remains challenging for the mechanistic research on new upconversion strategy of lanthanides. Here, a novel and efficient strategy to realize photon upconversion from more lanthanides and fine control of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service