Skip to Content
Merck
All Photos(1)

Key Documents

296937

Sigma-Aldrich

Di-tert-butylchlorosilane

97%

Synonym(s):

Chloro-di-tert-butylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2SiHCl
CAS Number:
Molecular Weight:
178.77
Beilstein:
2203045
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.441 (lit.)

bp

82-84 °C/45 mmHg (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[SiH](Cl)C(C)(C)C

InChI

1S/C8H19ClSi/c1-7(2,3)10(9)8(4,5)6/h10H,1-6H3

InChI key

OGWXFZNXPZTBST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 2927-2927 (1993)
Organometallics, 11, 3542-3542 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service