Skip to Content
Merck
All Photos(1)

Key Documents

278084

Sigma-Aldrich

3-Nitrophthalhydrazide

98%

Synonym(s):

2,3-Dihydro-5-nitro-1,4-phthalazinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5N3O4
CAS Number:
Molecular Weight:
207.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

>300 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1cccc2C(=O)NNC(=O)c12

InChI

1S/C8H5N3O4/c12-7-4-2-1-3-5(11(14)15)6(4)8(13)10-9-7/h1-3H,(H,9,12)(H,10,13)

InChI key

YVOBBFQJMOGQOU-UHFFFAOYSA-N

Related Categories

Application

3-Nitrophthalhydrazide has been used in the preparation of 3-aminophthalhydrazidate, required for the synthesis of monoacylhydrazidate-coordinated Mn2+ and Pb2+ compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Jin et al.
Dalton transactions (Cambridge, England : 2003), 41(20), 6137-6147 (2012-04-13)
By the simple hydrothermal self-assembly, four new monoacylhydrazidate-coordinated compounds [Mn(APTH)(2)(H(2)O)] 1, [Pb(APTH)(2)]·0.25H(2)O 2, [Pb(2)(EPDH)(4)(H(2)O)] 3 and [Pb(MPDH)(2)] 4 (APTH = 3-aminophthalhydrazidate, EPDH = 5-ethylpyridine-2,3-dicarboxylhydrazidate, MPDH = 6-methylpyrinde-2,3-dicarboxylhydrazidate) were obtained. It is noteworthy that APTH was derived from the in situ
Bin Li et al.
Analytical chemistry, 91(13), 8221-8228 (2019-06-01)
In many aspects of the matrix-assisted laser desorption/ionization mass spectrometry imaging (MALDI MSI) technique, the discovery of new MALDI matrixes has been a major task for the improvement of ionization efficiency, signal intensity, and molecular coverage. In this work, five

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service