Skip to Content
Merck
All Photos(1)

Key Documents

265314

Sigma-Aldrich

2,3-Difluorobenzyl bromide

98%

Synonym(s):

α-Bromo-2,3-difluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2Br
CAS Number:
Molecular Weight:
207.02
Beilstein:
7089244
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.528 (lit.)

density

1.628 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Fc1cccc(CBr)c1F

InChI

1S/C7H5BrF2/c8-4-5-2-1-3-6(9)7(5)10/h1-3H,4H2

InChI key

FTBSGSZZESQDBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3-Difluorobenzyl bromide was used in the synthesis of chemokine antagonists.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iain Walters et al.
Bioorganic & medicinal chemistry letters, 18(2), 798-803 (2008-02-02)
The CXCR2 SAR of a series of bicyclic antagonists such as the 2-aminothiazolo[4,5-d]pyrimidine 3b was investigated by systematic variation of the fused pyrimidine-based heterocyclic cores. Replacement of the aminothiazole ring with a 2-thiazolone alternative led to a series of thiazolo[4,5-d]pyrimidine-2(3H)-one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service