Skip to Content
Merck
All Photos(1)

Key Documents

248983

Sigma-Aldrich

(S)-(+)-α-Methoxyphenylacetic acid

99%

Synonym(s):

O-Methyl-L-mandelic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
2361718
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]17/D +150°, c = 1 in ethanol

mp

64-66 °C (lit.)

SMILES string

CO[C@H](C(O)=O)c1ccccc1

InChI

1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)/t8-/m0/s1

InChI key

DIWVBIXQCNRCFE-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in the synthesis of biologically active molecules including:
  • Acyclonucleoside phosphonates structural analogs of adefovir
  • Labeled discodermolide for studying binding to tubulin
  • 10-Isocyano-4-cadinene used for antifouling activity

Reactant involved in studies of immunostimulating chromanones gonytolides A-C

Reactant involved in:
  • Hydroxylations and epoxidations
  • Hydrogenations

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service