Skip to Content
Merck
All Photos(2)

Key Documents

192600

Sigma-Aldrich

2-Aminophenethyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2CH2OH
CAS Number:
Molecular Weight:
137.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.588 (lit.)

bp

147-148 °C/3.5 mmHg (lit.)

density

1.045 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

Nc1ccccc1CCO

InChI

1S/C8H11NO/c9-8-4-2-1-3-7(8)5-6-10/h1-4,10H,5-6,9H2

InChI key

ILDXSRFKXABMHH-UHFFFAOYSA-N

General description

2-Aminophenethyl alcohol undergoes one-pot cyclization with carboxylic acids in the presence of PPh3, CCl4 and NEt3 to yield N-acyl indolines.

Application

2-Aminophenethyl alcohol was used in the synthesis of:
  • indole derivatives
  • N-(cyanothioformyl)indoline
  • dihydro-3,1-benzoxazepine

Other Notes

This material may darken over time with minimal impact to chemical purity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

N-(Cyanothioformyl) indoline; a new indoline ring forming reaction.
Besson T, et al.
Journal of the Chemical Society. Perkin Transactions 1, 24, 4057-4060 (1998)
Zengxue Wang et al.
The Journal of organic chemistry, 72(24), 9364-9367 (2007-11-02)
A unique one-pot cyclization of 2-aminophenethyl alcohols with carboxylic acids in the presence of PPh3, CCl4, and NEt3 furnished the formation of N-acyl indolines in good to excellent yields. This new approach provides an efficient, scalable, low-cost, and direct access
Ruthenium-catalyzed dehydrogenative N-heterocyclization. Indoles from 2-aminophenethyl alcohols and 2-nitrophenethyl alcohols.
Tsuji Y, et al.
The Journal of Organic Chemistry, 55(2), 580-584 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service