Skip to Content
Merck
All Photos(2)

Key Documents

140465

Sigma-Aldrich

Di-tert-butyl hydrazodiformate

97%

Synonym(s):

tert-Butyl hydrazodiformate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NHCO2C(CH3)3]2
CAS Number:
Molecular Weight:
232.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

123-126 °C (dec.) (lit.)

functional group

amine

SMILES string

CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C

InChI

1S/C10H20N2O4/c1-9(2,3)15-7(13)11-12-8(14)16-10(4,5)6/h1-6H3,(H,11,13)(H,12,14)

InChI key

TYSZETYVESRFNT-UHFFFAOYSA-N

General description

Di-tert-butyl hydrazodiformate acts as hydrazine substrate during the preparation of protected pyridylhydrazine derivatives.

Application

Di-tert-butyl hydrazodiformate was used as starting reagent during the four-step synthesis of the aminobicyclopyrazolone hydrochloride. It was used as reagent during Pd-catalyzed amination reactions of halo-pyridine substrates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Convenient Synthesis of Pyrazolidine and 3-Amino-6, 7-dihydro-1H, 5H-pyrazolo [1, 2-a] pyrazol-1-one.
Boros EE, et al.
Journal of Heterocyclic Chemistry, 38(3), 613-616 (2001)
J B Arterburn et al.
Organic letters, 3(9), 1351-1354 (2001-05-12)
[reaction in text] Protected pyridylhydrazine derivatives were prepared in a one-step palladium-catalyzed amination reaction using chelating phosphine ligands. 2-Pyridyl chlorides, bromides, and triflates were effective electrophiles in these reactions. Di-tert-butyl hydrazodiformate was an excellent hydrazine substrate, and the resulting products
Jeffrey B Arterburn et al.
The Journal of organic chemistry, 68(18), 7063-7070 (2003-08-30)
The development of (99m)Tc-estradiol radiopharmaceuticals would be advantageous for the detection of estrogen receptor-positive breast tumors. Estradiol derivatives conjugated to organometallic tricarbonyl-Tc(I) and related Re(I) complexes are capable of achieving high receptor binding affinity, but effective methods for synthesizing radiolabeled
Daniel T Cohen et al.
Cell reports, 30(10), 3229-3239 (2020-03-12)
BCL-2 family proteins converge at the mitochondrial outer membrane to regulate apoptosis and maintain the critical balance between cellular life and death. This physiologic process is essential to organism homeostasis and relies on protein-protein and protein-lipid interactions among BCL-2 family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service