Merck
Ebselen, a Small-Molecule Capsid Inhibitor of HIV-1 Replication.
Antimicrobial agents and chemotherapy (2016-01-27)
Suzie Thenin-Houssier, Ian Mitchelle S de Vera, Laura Pedro-Rosa, Angela Brady, Audrey Richard, Briana Konnick, Silvana Opp, Cindy Buffone, Jakob Fuhrmann, Smitha Kota, Blase Billack, Magdalena Pietka-Ottlik, Timothy Tellinghuisen, Hyeryun Choe, Timothy Spicer, Louis Scampavia, Felipe Diaz-Griffero, Douglas J Kojetin, Susana T Valente
ABSTRACT
The human immunodeficiency virus type 1 (HIV-1) capsid plays crucial roles in HIV-1 replication and thus represents an excellent drug target. We developed a high-throughput screening method based on a time-resolved fluorescence resonance energy transfer (HTS-TR-FRET) assay, using the C-terminal domain (CTD) of HIV-1 capsid to identify inhibitors of capsid dimerization. This assay was used to screen a library of pharmacologically active compounds, composed of 1,280in vivo-active drugs, and identified ebselen [2-phenyl-1,2-benzisoselenazol-3(2H)-one], an organoselenium compound, as an inhibitor of HIV-1 capsid CTD dimerization. Nuclear magnetic resonance (NMR) spectroscopic analysis confirmed the direct interaction of ebselen with the HIV-1 capsid CTD and dimer dissociation when ebselen is in 2-fold molar excess. Electrospray ionization mass spectrometry revealed that ebselen covalently binds the HIV-1 capsid CTD, likely via a selenylsulfide linkage with Cys198 and Cys218. This compound presents anti-HIV activity in single and multiple rounds of infection in permissive cell lines as well as in primary peripheral blood mononuclear cells. Ebselen inhibits early viral postentry events of the HIV-1 life cycle by impairing the incoming capsid uncoating process. This compound also blocks infection of other retroviruses, such as Moloney murine leukemia virus and simian immunodeficiency virus, but displays no inhibitory activity against hepatitis C and influenza viruses. This study reports the use of TR-FRET screening to successfully identify a novel capsid inhibitor, ebselen, validating HIV-1 capsid as a promising target for drug development.
Sigma-Aldrich
Ebselen Oxide, ≥98% (HPLC)