Skip to Content
Merck
Pyrogallol Red Suitable for use as a complexometric indicator

Pyrogallol Red

Pyrogallol Red Suitable for use as a complexometric indicator
Synonym(s):
Pyrogallolsulfonephthalein
Empirical Formula (Hill Notation):
C19H12O8S
CAS Number:
Molecular Weight:
400.36
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing