Skip to Content
Merck
Campylobacter Culture Medium powder, final pH 7.3±0.2 (25 °C), bottle of 500 g, non-sterile, suitable for microbiology

Campylobacter Culture Medium

Campylobacter Culture Medium powder, final pH 7.3±0.2 (25 °C), bottle of 500 g, non-sterile, suitable for microbiology

Products

Product Category
Brand
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility