21378
Campylobacter Culture Medium
powder, final pH 7.3±0.2 (25 °C), bottle of 500 g, non-sterile, suitable for microbiology
Synonym(s):
Campylobacter Selective Agar (Base)
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
agar, 13 g/L
electrolyte, 5 g/L
peptone-protein mixture, 21 g/L
starch (soluble), 1 g/L
packaging
bottle of 500 g
final pH
7.3±0.2 (25 °C)
application(s)
clinical testing
environmental
food and beverages
veterinary
microbiology
suitability
selective for Campylobacter spp.
Looking for similar products? Visit Product Comparison Guide
Application
Preparation Note
50-70 ml/l defibrinated blood
1 vl/l Park and Sanders Selective Supplement I (Cat. No. 17191)
1 vl/l Park and Sanders Selective Supplement II (Cat. No. 17194)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service