Skip to Content
Merck
All Photos(9)

Documents

ZRB2337

Sigma-Aldrich

Anti-MGEA5/OGA Antibody, clone 1B3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Beta-N-acetylglucosaminidase, Beta-N-acetylhexosaminidase, Beta-hexosaminidase, EC:3.2.1.169, Meningioma-expressed antigen 5, N-acetyl-beta-D-glucosaminidase, N-acetyl-beta-glucosaminidase, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, NCOAT, Protein O-GlcNAcase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B3, recombinant monoclonal

description

1B3 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 102.92 kDa
observed mol wt ~120 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey, horse, canine, porcine, bovine, feline

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... OGA(10724)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B3 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects O-GlcNAcase (OGA/MGEA5). It targets an epitope within 24 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the internal region of human O-GlcNAcase.

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MGEA5/OGA in NIH3T3 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected MGEA5/OGA in HeLa and HepG2 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected MGEA5/OGA in Human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound MGEA5/OGA peptide with a KD of 9.9 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein O-GlcNAcase (UniProt: O60502; also known as EC:3.2.1.169, OGA, Beta-N-acetylglucosaminidase, Beta-N-acetylhexosaminidase, Beta-hexosaminidase, Meningioma-expressed antigen 5, N-acetyl-beta-D-glucosaminidase, N-acetyl-beta-glucosaminidase, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, NCOAT) is encoded by the OGA (also known as HEXC, KIAA0679 MEA5, MGEA5) gene (Gene ID: 10724) in human. O-GlcNAcylation is an important posttranslational modification found on serine and threonine residues that is brought about by O-GlcNAc transferase (OGT) and O-GlcNAcase (OGA). They mediate the dynamic cycling of O-GlcNAcylation on a wide variety of cytosolic, nuclear, and mitochondrial proteins. In this cycling, OGT catalyzes the addition of O-GlcNAc to proteins and OGA catalyzes the removal of this modification. Disruption in the activity of OGT and OGA has been implicated in multiple disease states. Deletion of genes for these enzymes causes early postnatal lethality. OGA is an evolutionarily conserved,  monomeric enzyme that is ubiquitously distributed. Its highest expression is observed in the brain, placenta, and pancreas. Four isoforms of OGA have been described that are produced by alternative splicing. Isoform 1 (long isoform)  is reported to cleave GlcNAc but nor GalNAc from O-glycosylated proteins. Its activity is inhibited by N-acetylglucosamine and not N-acetylgalactosamine. The long isoform of OGA resides mainly within the cytosol, whereas the short isoform localizes in the nucleus and lipid droplets, and it has lower enzymatic activity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ong, Q., et al. (2018). Front. Endocrinol. 9; 599; Bond, MR., and Hanover, JA. (2013). Annu. Rev. Nutr. 33; 205-29; Yang, YR.,  et al. (2012). Aging Cell. 11(3); 439-448; Gao, Y., et al. (2001). J. Biol. Chem. 276(13); 9838-9845).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service