Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1984

Sigma-Aldrich

Anti-Phospho-NDRG1 (Thr346) Antibody, clone 5N4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein NDRG1;Differentiation-related gene 1 protein;DRG-1;N-myc downstream-regulated gene 1 protein;Nickel-specific induction protein Cap43;Reducing agents and tunicamycin-responsive protein;RTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5N4, recombinant monoclonal

description

5N4 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 42.84 kDa
observed mol wt ~45 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

monkey, rat, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pThr346)

Gene Information

human ... NDRG1(10397)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5N4 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects NDRG1 phosphorylated on Threonine 346.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphothreonine 346 of human NDRG1.

Application

Quality Control Testing

Evaluated by Western Blotting in U87 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Phospho-NDRG1 (Thr346) in U87 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Phospho-NDRG1 (Thr346) in NIH3T3 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound Phospho-NDRG1 (Thr346) peptide with a KD of 6.9 x 10-6 in an affinity binding assay.

Peptide Inhibition Assay: Target band detection in lysate from U87 cells was prevented by pre-blocking of a representative lot with phosphorylated NDRG1 (Thr346) peptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Phospho-NDRG1 (Thr346) in C2C12 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Phospho-NDRG1 (Thr346) in human colon and human cervical cancer tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Protein NDRG1 (UniProt: Q92597; also known as Differentiation-related gene 1 protein, DRG-1, N-myc downstream-regulated gene 1 protein, Nickel-specific induction protein Cap43, Reducing agents and tunicamycin-responsive protein, RTP) is encoded by the NDRG1 (also known as CAP43, DRG1, RTP) gene (Gene ID: 10397) in human. NDRG1 is a stress-responsive protein that is induced under a wide variety of stress and cell growth-regulatory conditions. It is ubiquitously expressed but the most prominent expression is observed in placental membranes, prostate, kidney, small intestine, and ovary tissues. It is mainly a cytoplasmic protein, but is differentially localized to other regions. NDRG1 is reported to be up-regulated by cell differentiation signals in various cancer cell lines and also suppresses tumor metastasis. It is reported to play different roles in metastatic versus non-metastatic cells. It suppresses cell growth and proliferation in the metastatic H1299-NDRG1 cells, but does not have any effect on the non-metastatic DLD-1-NDRG1 cells. It is induced about 20-fold during in vitro differentiation of the colon carcinoma cell lines HT-29-D4 and Caco-2 cells. NDRG1 is shown to be essential, but not sufficient for p53/TP53-mediated caspase activation and apoptosis. It is also shown to have a role in cell trafficking of Schwann cells and is necessary for the maintenance and development of the peripheral nerve myelin sheath. NDRG1 is phosphorylated by several protein kinases, including PKA, PKC, and CaMK II. It is also phosphorylated by SGK1 on many serine and threonine residues in the C-terminal region, which is temporally and spatially controlled during the cell cycle. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.(Ref.: (Ref.: McCaig, C., et al. (2011). Biochem. Biophys. Res. Commun. 411(2); 227-234; Ellen, TP., et al. (2008). Carcinogenesis. 29(1); 2-8).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service