Z740236
TLC Plastic Sheets
L × W 20 cm × 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent indicator: no
Synonym(s):
Thin Layer Chromatography Sheets
About This Item
Recommended Products
material
plastic support
silica gel 60 matrix
Quality Level
feature
binder Polymeric
fluorescent indicator: no
packaging
pkg of 25 ea
manufacturer/tradename
EMD Millipore® 1.05748.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
200 μm
particle size
10-12 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service