Key Documents
Z663336
Brady® thermal labeling system TLSPCLink printers
labeling system with Labelmark software
About This Item
UNSPSC Code:
55121611
manufacturer/tradename
Brady
availability
available only in EU
General description
Printer system includes Labelmark software, battery pack, AC adapter/battery charger, Resin black ribbon, sample labels, manual, Quick Start Guide, communications cables and Windows drivers.
The TLS Thermal Labeling System delivers permanent, quick and clear printing, label after label. With thermal transfer printing there is no need to handle your materials delicately, there′s no smearing!
The system automatically recognises label sizes and printer settings and has a built-in cutter, eliminating wasted time and materials.
The TLS PC Link is the ideal economical choice if you have no need for a portable solution and you prepare your labeling projects at your desk.
  • Compact professional desktop labeling system
  • Includes Labelmark software
  • Can be used with any Windows based software
  • Printer control language can be integrated with non-windows based equipment
  • AC power supply automatically charges battery
  • Print length up to 152.4 mm
Legal Information
Brady is a registered trademark of Brady Worldwide, Inc.