Z640387
Eppendorf® epT.I.P.S. Reloads
volume range 0.1-10 μL
Synonym(s):
epT.I.P.S. Reload System, pipetting tips
About This Item
Eppendorf® 022491504
Recommended Products
material
dark gray tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030 073.363
Eppendorf® 022491504
L
34 mm
maximum volume
10 μL
volume range
0.1-10 μL
volume
0.1—10 μL
Looking for similar products? Visit Product Comparison Guide
General description
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately)
Application
Features and Benefits
- Less waste compared to disposable racks
- Autoclavable refill trays without foil, for reuse
- Refill system packaged as dual-sided or in stack form, depending on tip size
- Color-coded trays for easy volume and pipette identification
- Single-use pipette tips
- Universal cone for compatibility
- Easy to handle
- Volume marking for visual inspection
- Easy attachment and ejection with minimal force
- Provides complete sealing
- Ensures maximum precision and accuracy
- Long pipette tips available for certain volume sizes
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service