Skip to Content
Merck
All Photos(5)

Key Documents

WH0006748M1

Sigma-Aldrich

Monoclonal Anti-SSR4 antibody produced in mouse

clone 2D3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TRAPD, Anti-signal sequence receptor, delta (translocon-associated protein delta)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SSR4(6748)

Related Categories

General description

SSR4, also called TRAPD, is assumed to be involved in protein secretion. It is located in the Xq28 region, arranged in a compact head-to-head manner with the IDH3G gene. These two genes are driven by a bidirectional promoter located between them, and encode proteins involved in unrelated biochemical pathways located in different compartments of the cell. The nontranscribed intergenic region represents only 133 bp and is embedded in a CpG island. The CpG island functions as a bidirectional promoter to initiate the transcription of both functionally unrelated genes with distinct expression patterns. SSR4 consists of six exons and is approximately 70 kb telomeric to the ALD gene. Although alternative splicing of exon 5 has not been detected in human SSR4, transcript variants missing the region homologous to human exon 5 have been detected in both Xenopus laevis and Mus musculus. (provided by RefSeq)

Immunogen

SSR4 (AAH03371, 24 a.a. ~ 173 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EACLEPQITPSYYTTSDAVISTETVLIVEISLTCKNRVQNMALYADVGGKQFPVTRGQDVGRYQVSWSLDHKSAHAGTYEVRFFDEESYSLLRKAQRNNEDISIIPPLFTVSVDHRGTWNGPWVSTEVLAAAIGLVIYYLAFSAKSHIQA

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Estelle Losfeld et al.
Human molecular genetics, 23(6), 1602-1605 (2013-11-13)
Nearly 50 congenital disorders of glycosylation (CDG) are known, but many patients biochemically diagnosed with CDG do not have mutations in known genes. Here, we describe a 16-year-old male who was born with microcephaly, developed intellectual disability, gastroesophageal reflux and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service