Skip to Content
Merck
All Photos(1)

Key Documents

SYNF81221LN

Synthware Volumetric flask, Class A with glass stopper

borosilicate glass 3.3, capacity 1000 mL

Synonym(s):

Synthware F81221LN, measuring flask, graduated flask

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.43

material

PTFE
borosilicate glass 3.3

feature

class A

packaging

pack of 1 ea

manufacturer/tradename

Kemtech F81221LN

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Volumetric Flask,Heavy Wall,1000mL,Class A,#22 glass Stopper

Application

kemetch; synthware; flask; volumetric flask; class a

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service