Skip to Content
Merck
All Photos(2)

Key Documents

SRP5251

Sigma-Aldrich

PKC-α, active, GST tagged from Xanopus sp.

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC82897, PRKCA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

Xenopus sp.

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

186-290 nmol/min·mg

mol wt

~103 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

Xenopus laevis ... prkca(447031)

General description

Protein kinase C-α (PKC-α) is a member of the PKC family of serine- and threonine-specific protein kinases that can be activated by calcium and the secondary messenger diacylglycerol. The protein is expressed in the dorsal ectoderm of Xenopus.

Biochem/physiol Actions

Protein kinase C-α (PKC-α) has been reported to play roles in many different cellular processes, such as cell adhesion, cell transformation, cell cycle checkpoint and cell volume control. It has been identified as a fundamental regulator of cardiac contractility and Ca2+ handling in myocytes.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

aPKC phosphorylates p27Xic1, providing a mechanistic link between apicobasal polarity and cell-cycle control.
Sabherwal N
Developmental Cell, 31(5), 559-571 (2014)
Protein kinase C isozymes have distinct roles in neural induction and competence in Xenopus.
Otte AP and Moon RT
Cell, 68(6), 1021-1029 (1992)
PKC-alpha regulates cardiac contractility and propensity toward heart failure.
Braz JC
Nature Medicine, 10(3), 248-254 (2004)
Yi Liu et al.
The Biochemical journal, 478(10), 1999-2017 (2021-05-08)
Human hepatic tryptophan 2,3-dioxygenase (hTDO) is a homotetrameric hemoprotein. It is one of the most rapidly degraded liver proteins with a half-life (t1/2) of ∼2.3 h, relative to an average t1/2 of ∼2-3 days for total liver protein. The molecular mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service