Skip to Content
Merck
All Photos(1)

Documents

SRP2075

Sigma-Aldrich

SRC1, FLAG tagged, TEV site human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

F-SRC-1, KAT13A, MGC129719, MGC129720, RIP160, SRC1, bHLHe42, bHLHe74

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~19.5 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

1000 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NCOA1(8648)

Biochem/physiol Actions

Steroid receptor coactivator 1 (SRC1) is a transcriptional coactivator that mediates the activating functions of many of the nuclear hormone receptors. It is also known as NCoA1 and is a member of the SRC/p160 coactivator family. SRC1 has been shown to be over expressed in some cancers. SRC1 is a 160 kDa protein that contains several LXXLL motifs, which are involved in nuclear receptor interaction. The region 627-786 contains 3 LXXLL motifs that are involved in interaction with nuclear hormone receptorsand have been previously used in assays detecting ligand-dependent receptor-cofactor interactions. This protein contains the Tobacco Etch virus (TEV) protease site and can be cleaved using acTEV to yield untagged SRC1 RID. This protein will be useful for assays where a native untagged protein is desired.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Oñate et al.
Science (New York, N.Y.), 270(5240), 1354-1357 (1995-11-24)
A yeast two-hybrid system was used to identify a protein that interacts with and enhances the human progesterone receptor (hPR) transcriptional activity without altering the basal activity of the promoter. Because the protein stimulated transactivation of all the steroid receptors
Toshihiko Yanase et al.
Internal medicine (Tokyo, Japan), 43(5), 368-373 (2004-06-23)
Coregulators are a group of proteins, which modulate the nuclear receptor transactivation function. In this study, a new "coregulator disease" concept was proposed from observations of a case of androgen insensitivity syndrome (AIS) and cases involving Rubinstein-Taybi syndrome and X-linked
D M Heery et al.
Nature, 387(6634), 733-736 (1997-06-12)
The binding of lipophilic hormones, retinoids and vitamins to members of the nuclear-receptor superfamily modifies the DNA-binding and transcriptional properties of these receptors, resulting in the activation or repression of target genes. Ligand binding induces conformational changes in nuclear receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service