Skip to Content
Merck
All Photos(1)

Documents

SML4019

Sigma-Aldrich

Neuromedin U-23 (rat) trifluoroacetate

new

≥95% (HPLC)

Synonym(s):

L-Tyrosyl-L-lysyl-L-valyl-L-asparaginyl-L-α-glutamyl-L-tyrosyl-L-glutaminylglycyl-L-prolyl-L-valyl-L-alanyl-L-prolyl-L-serylglycylglycyl-L-phenylalanyl-L-phenylalanyl-L-leucyl-L-phenylalanyl-L-arginyl-L-prolyl-L-arginyl-L-aspartamide trifluoroacetate, Neuromedin U trifluoroacetate, Phe-Lys-Ala-Glu-Tyr-Gln-Ser-Pro-Ser-Val-Gly-Gln-Ser-Lys-Gly-Tyr-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2 trifluoroacetate, Rat neuromedin U-23 trifluoroacetate, YKVNEYQGPVAPSGGFFLFRPRN-NH2 trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C124H180N34O31
Molecular Weight:
2642.97
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

O=C([C@H](CC1=CC=CC=C1)NC([C@H](CC(C)C)NC([C@@H](NC([C@@H](NC(CNC(CNC([C@H](CO)NC([C@H]2CCCN2C([C@@H](NC([C@H](C(C)C)NC(C3N(CCC3)C(CNC([C@@H](NC([C@H](CC4=CC=C(C=C4)O)NC([C@@H](NC([C@@H](NC([C@@H](NC([C@H](CCCCN)NC([C@@H](N)CC5=CC=C(C=C5)O)=O)=O)C(C)C)=O)

Biochem/physiol Actions

Endogenous agonist of neuromedin-U receptor 1 (NMUR1) and neuromedin-U receptor 2 (NMUR2)



Neuromedin U-23 (Neuromedin U), a neuropeptide, is endogenous agonist of neuromedin-U receptor 1 (EC50 = 0.17 nM) and neuromedin-U receptor 2 (EC50 = 1.4 nM) that regulates blood pressure, ion transport in the gut, mesenteric blood flow and adrenocortical function. Neuromedin U-23 decreases body weight and food intake and increases core body temperature in mice when administered at a dose of 36 µg/animal. It appears that neuromedin U regulates epithelial cell differentiation and barrier immunity through interaction with neuromedin-U receptor 1 present at small intestine eosinophils.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Peier et al.
Endocrinology, 150(7), 3101-3109 (2009-03-28)
Neuromedin U (NMU) and neuromedin S (NMS) are structurally related neuropeptides that have been reported to modulate energy homeostasis. Pharmacological data have shown that NMU and NMS inhibit food intake when administered centrally and that NMU increases energy expenditure. Additionally
Yu Li et al.
Science (New York, N.Y.), 381(6663), 1189-1196 (2023-09-14)
Eosinophils are granulocytes that play an essential role in type 2 immunity and regulate multiple homeostatic processes in the small intestine (SI). However, the signals that regulate eosinophil activity in the SI at steady state remain poorly understood. Through transcriptome
Luca Botticelli et al.
Pharmacological research, 195, 106875-106875 (2023-07-31)
Neuromedin U (NMU) is a bioactive peptide produced in the gut and in the brain, with a role in multiple physiological processes. NMU acts by binding and activating two G protein coupled receptors (GPCR), the NMU receptor 1 (NMU-R1), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service