Skip to Content
Merck
All Photos(1)

Key Documents

SML2880

Sigma-Aldrich

SGC-CLK-1

≥98% (HPLC)

Synonym(s):

N-(3-Methoxy-5-(trifluoromethyl)phenyl)-4-(6-methoxypyrazolo[1,5-b]pyridazin-3-yl)pyrimidin-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15F3N6O2
CAS Number:
Molecular Weight:
416.36
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

FC(F)(F)C1=CC(OC)=CC(NC2=NC=CC(C3=C(C=C4)N(N=C3)N=C4OC)=N2)=C1

InChI

1S/C19H15F3N6O2/c1-29-13-8-11(19(20,21)22)7-12(9-13)25-18-23-6-5-15(26-18)14-10-24-28-16(14)3-4-17(27-28)30-2/h3-10H,1-2H3,(H,23,25,26)

InChI key

GJYVLTPTDBQQCY-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective CLK1/2/4 inhibitor with good kinome-wide selectivity
SGC-CLK-1 is a potent and selective CLK1/2/4 inhibitor (IC50 = 13/4/46 nM by enzyme assay; IC50 = 165/70/100 nM by cell-based NanoBRET assay) with good kinome-wide selectivity (by 403-panel KINOMEscan). SGC-CLK-1 inhibits CLK3 at significantly higher concentrations (IC50 = 363 nM by enzyme assay) and causes only partial STK16 inhibition by cell-based NanoBRET assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SGC-CLK-1 (CAF-170) a chemical probe for the Cdc2-Like kinases CLK1, CLK2, and CLK4
bioRxiv : the preprint server for biology (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service