Skip to Content
Merck
All Photos(1)

Key Documents

SML0558

Sigma-Aldrich

BETP

≥98% (HPLC)

Synonym(s):

2-(Ethylsulfinyl)-4-[3-(phenylmethoxy)phenyl]-6-(trifluoromethyl)-pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17F3N2O2S
CAS Number:
Molecular Weight:
406.42
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CCS(=O)c1nc(cc(n1)C(F)(F)F)-c2cccc(OCc3ccccc3)c2

InChI

1S/C20H17F3N2O2S/c1-2-28(26)19-24-17(12-18(25-19)20(21,22)23)15-9-6-10-16(11-15)27-13-14-7-4-3-5-8-14/h3-12H,2,13H2,1H3

InChI key

NTDFYGSSDDMNHI-UHFFFAOYSA-N

Biochem/physiol Actions

BETP is a GLP-1 receptor allosteric modulator.
BETP is a positive allosteric modulator of the GLP-1 receptor that potentiates cAMP accumulation and glucose-dependent insulin release in pancreatic islet cells, in the presence of the weak GLP-1 metabolite, GLP-1(9-36). BETP has additive effects on the endogenous GLP-1 receptor ligand GLP-1(7-36).
BetP is a trimeric Na+-coupled betaine symporter. BetP belongs to the betaine-choline-carnitine transporter (BCCT) family. It controls transport activity in dependence of the cytoplasmic K+-concentration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid-protein interactions in the regulated betaine symporter BetP probed by infrared spectroscopy.
Guler G, et al.
The Journal of Biological Chemistry, 291(9), 4295-4307 (2016)
Locating an extracellular K+-dependent interaction site that modulates betaine-binding of the Na+-coupled betaine symporter BetP.
Ge L, et al.
Proceedings of the National Academy of Sciences of the USA (2011)
Khyati Girdhar et al.
Journal of biomolecular structure & dynamics, 37(15), 3976-3986 (2018-10-10)
The glucagon-like peptide-1 receptor (GLP-1R) is a well-known target of therapeutics industries for the treatment of various metabolic diseases like type 2 diabetes and obesity. The structural-functional relationships of small molecule agonists and GLP-1R are yet to be understood. Therefore
C Weston et al.
British journal of pharmacology, 171(15), 3651-3665 (2014-04-10)
The glucagon-like peptide 1 (GLP-1) receptor performs an important role in glycaemic control, stimulating the release of insulin. It is an attractive target for treating type 2 diabetes. Recently, several reports of adverse side effects following prolonged use of GLP-1
Daniela G Dengler et al.
SLAS discovery : advancing life sciences R & D, 26(1), 1-16 (2020-08-05)
The secretin receptor (SCTR), a prototypical class B G protein-coupled receptor (GPCR), exerts its effects mainly by activating Gαs proteins upon binding of its endogenous peptide ligand secretin. SCTRs can be found in a variety of tissues and organs across

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service