Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300599

Sigma-Aldrich

Anti-MET (Ab-1003) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AUTS9 antibody produced in rabbit, Anti-HGFR antibody produced in rabbit, Anti-RCCP2 antibody produced in rabbit, Anti-c-Met antibody produced in rabbit, Anti-met proto-oncogene (hepatocyte growth factor receptor) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~156 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(V-D-Y-R-A)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MET(4233)

General description

MET/c-Met (mesenchymal-epithelial transition factor) is a tyrosine kinase receptor , that is located on chromosome 7q31.

Immunogen

Peptide sequence around aa. 1001-1005 (V-D-Y-R-A), according to the protein MET.

Biochem/physiol Actions

MET/c-Met (mesenchymal-epithelial transition factor) controls several biological processes like cell scattering, survival, proliferation, migration and c-MET signaling.(31)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for hepatocyte growth factor and scatter factor. Has a tyrosine-protein kinase activity. Functions in cell proliferation, scattering, morphogenesis and survival.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuo Hayashi et al.
Histopathology, 78(7), 987-999 (2020-11-30)
In the evolving era of precision medicine, increasing emphasis is placed on detecting molecular alterations driving the development of specific cancers and targeting them with matched therapies that can yield the best outcomes for patients. Lung adenocarcinomas with uncommon actionable
The c-Met receptor: Implication for targeted therapies in colorectal cancer
Safaie Qamsari E, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(5) (2017)
Nayeon Lee et al.
Biochemistry and biophysics reports, 26, 100973-100973 (2021-03-16)
Hepatocyte growth factor (HGF) is a neurotrophic factor and its role in peripheral nerves has been relatively unknown. In this study, biological functions of HGF and its receptor c-met have been investigated in the context of regeneration of damaged peripheral
Kyeong Ryang Ko et al.
Scientific reports, 8(1), 8316-8316 (2018-05-31)
During the peripheral nerve regeneration process, a variety of neurotrophic factors play roles in nerve repair by acting on neuronal or non-neuronal cells. In this report, we investigated the role(s) of hepatocyte growth factor (HGF) and its receptor, c-met, in
Phase II and biomarker study of the dual MET/VEGFR2 inhibitor foretinib in patients with papillary renal cell carcinoma
Choueiri TK, et al.
Journal of Clinical Oncology, 31(2), 181-186 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service