Skip to Content
Merck
All Photos(1)

Key Documents

SAB2502005

Sigma-Aldrich

Anti-IL7R antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CD127, IL-7R-alpha, IL7RA, ILRA, interleukin 7 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... IL7R(16197)

Immunogen

Peptide with sequence DDHSFWCHSQLE, from the internal region of the protein sequence according to NP_032398.3

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sae-Kyung Lee et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(13), 3485-3495 (2014-05-07)
The low immunogenicity of many cancer cells and the immunosuppression by various cancers and anticancer therapies have been an obstacle in the development of efficacious immunotherapies. Our goal was to test whether Toll-like receptor (TLR) agonists and anticancer chemotherapeutic agents
J Marius Munneke et al.
Blood, 124(5), 812-821 (2014-05-24)
Allogeneic hematopoietic stem cell transplantation (HSCT) is widely used to treat hematopoietic cell disorders but is often complicated by graft-versus-host disease (GVHD), which causes severe epithelial damage. Here we have investigated longitudinally the effects of induction chemotherapy, conditioning radiochemotherapy, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service