Skip to Content
Merck
All Photos(2)

Documents

PRS4809

Sigma-Aldrich

Anti-S1P1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ECGF1, Anti-EDG1, Anti-S1PR1, Anti-Sphingosine-1-phosphate receptor 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 42 kDa

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S1PR1(1901)

General description

S1P1 (sphingosine-1-phosphate receptor 1) is one of the transmembrane G protein coupled receptor. It is a member of the endothelial differentiation gene (EDG) family of receptors, hence it is also known as endothelial differentiation gene (EDG-1). It is located on chromosome 1p21.

Immunogen

a 14 amino acid peptide near the carboxy-terminus of the human S1P1.

Biochem/physiol Actions

S1P1 (sphingosine-1-phosphate receptor 1) acts as an anti-apoptotic/pro-proliferative protein in AML (acute myeloid leukemia) cells. It participates in angiogenesis and lymphangiogenesis. It is considered as a pro-tumorigenic receptor, as it induces migration of cancer cell and invasion along with tumor neovascularization.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4809.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingosine-1-phosph ate receptor 2
Adada M,et al.
FEBS Journal (2013)
S1PR1 mediates anti?apoptotic/pro?proliferative processes in human acute myeloid leukemia cells
Xu XQ, et al.
Molecular Medicine Reports, 14(4), 3369-3375 (2016)
Babita Rahar et al.
High altitude medicine & biology, 20(1), 78-88 (2019-03-21)
High altitude exposure alters biochemical, metabolic, and physiological features of heart and skeletal muscles, and hence has pathological consequences in these tissues. Central to these hypoxia-associated biochemical/metabolic shuffling are energy deficit accumulation of free radicals and ensuing oxidative damage in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service