MAK325
Cholesterol Uptake Assay Kit
sufficient for 100 fluorometric tests
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 fluorometric tests
detection method
fluorometric
relevant disease(s)
obesity; cardiovascular diseases; cancer
storage temp.
−20°C
Related Categories
General description
Cholesterol, a crucial sterol and lipid component, is present in cell membranes and circulates in the bloodstream of all animals. It plays essential roles in maintaining cell membrane structure, hormone synthesis, and facilitating cell signaling processes. The intricate regulation of cellular cholesterol levels is paramount, as imbalances have been linked to conditions such as obesity and heart disease. Moreover, heightened cholesterol uptake has been correlated with the rapid proliferation of cancer cells. Through monitoring cellular cholesterol uptake, researchers can delve into these pressing health concerns, advancing both drug discovery efforts and biochemical research aimed at identifying potential treatment modalities.
Application
The Cholesterol Uptake Assay Kit may be used for:
- Drug Discovery
- Cardiovascular Research
- Obesity Research
- Cancer Research
Features and Benefits
- Simplified Process: Experience a streamlined procedure as cell treatment occurs directly in the fluorescence 96-well plate, saving time and effort.
- High-Throughput Compatibility: Seamlessly integrate the kit into high-throughput handling systems for enhanced efficiency and accuracy.
Suitability
Suitable for testing cholesterol uptake by adherent cells, screening of
cholesterol uptake inhibitors, and evaluation of effect of drugs on
cholesterol uptake.
cholesterol uptake inhibitors, and evaluation of effect of drugs on
cholesterol uptake.
Principle
The cholesterol uptake assay kit is based on cellular uptake of a fluorescently tagged cholesterol probe. The fluorescence intensity measured at λex/em = 485/535 nm is proportional to the amount of cholesterol uptaken by the cells.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3
Target Organs
Central nervous system, Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service