Skip to Content
Merck
All Photos(1)

Key Documents

M042130

Sigma-Aldrich

DMT-Biotin-C6 Phosphoramidite

configured for ABI

Synonym(s):

5′ Biotin Amidite, 5′-Biotin Phosphoramidite, 6-[(2-Cyanoethoxy)diisopropylaminophosphanoxy]-[1-N-(4,4′-dimethoxytrityl)biotinyl]aminohexane, DMT-Biotin-C6 Amidite, N-DMT-Biotinyl-2-Aminoethoxyethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H64N5O6PS
CAS Number:
Molecular Weight:
846.07
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

product line

Proligo Reagents

Assay

≥96.0% (reversed phase HPLC)
≥97% (31P-NMR)

suitability

conforms to structure for H-NMR
conforms to structure for H-NMR

compatibility

configured for ABI

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(N2[C@H]3CS[C@@H](CCCCC(=O)NCCCCCCOP(OCCC#N)N(C(C)C)C(C)C)[C@H]3NC2=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C46H64N5O6PS/c1-34(2)51(35(3)4)58(57-32-16-29-47)56-31-15-8-7-14-30-48-43(52)20-13-12-19-42-44-41(33-59-42)50(45(53)49-44)46(36-17-10-9-11-18-36,37-21-25-39(54-5)26-22-37)38-23-27-40(55-6)28-24-38/h9-11,17-18,21-28,34-35,41-42,44H,7-8,12-16,19-20,30-33H2,1-6H3,(H,48,52)(H,49,53)/t41-,42-,44-,58?/m0/s1

InChI key

RBXYKQBFPLXSJS-RYQQVEDISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service